Single stage gas regulators
Supplier: Restek
Certificates
About this item
These stainless steel regulators are the standard for ultra high purity and corrosion resistant pressure regulation. They are more easily purged of atmospheric components compared to brass gas regulators.
- Use when there is secondary pressure regulation downstream
- Identical gas purity protection as with dual-stage gas regulators
- Especially useful in atmospheres of dry corrosive gases such as hydrogen
- Outlet assembly: 316 L stainless steel diaphragm valve and ¹/₄ tube fitting
Frequently Bought Together