Order Entry
Slovenia
ContactUsLinkComponent
Human Recombinant MetAP2 (from Insect cells)
Human Recombinant MetAP2 (from Insect cells)
Catalog # ENZOBMLSE5690010
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Human Recombinant MetAP2 (from Insect cells)
Catalog # ENZOBMLSE5690010
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Enzyme type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Seq:
    02
  • Environmentally Preferable:
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Enzyme name:
    MetAP2
  • Enzyme synonyms:
    Methionine aminopeptidase 2, p67eIF2, Eukaryotic initiation factor 2-associated protein
  • Purity:
    ≥95%
  • Molecular weight:
    ~70 kDa
  • pH:
    7,3

Specifications

About this item

MetAP2 is a metalloenzyme that catalyses removal of the N-terminal methionine from proteins, with the physiologically relevant metal thought to be either cobalt or manganese.

  • Produced in insect cells as recombinant, full-length (aa 1-478) methionine aminopeptidase 2 with an N-terminal hemagglutinin (HA) tag (YPYDVPDYA), cloned from human cDNA
  • Purified by chromatography (SDS-PAGE)
  • Highly active

It has a liquid formulation in 20 mM HEPES at pH 7,3, containing 0,1 M KCl, 1 mM CoCl2, 1,5 mM MgCl2, 0,02% sodium azide and 10% glycerol.