Disinfectant wipes, Bactinyl®
Supplier: ORAPI EUROPE
Certificates
About this item
Pre-saturated wipes for cleaning, pre-disinfection and disinfection of contact equipment, sensors and instruments. The wipes kill 99,999% of all germs targeted.
- Available perfumed or odourless
- Can be used on all types of materials
- Practical tub dispenser with lid
Bactericidal: NF EN 1040, NF EN 1276.
Fungicide: NF EN 1275, NF EN 1650.
Virucidal: NF T 72-180 on Rotavirus, HIV-1, Poliovirus, Herpes virus. NF EN 14476 on H1N1.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Related Information
Certificates
Search by Lot NumberBactinyl® Disinfectant Wipes
Product Literature
GARCLB920600_TDS_EN
Datasheet
Frequently Bought Together