Tubing cutting pliers
Catalog # REST20193
Supplier: Restek
Specifications
- Description:Tubing cutting pliers, for 1,6 mm tubing
- Pk:1
Specifications
About this item
1/16" tubing cutting pliers.
- Cuts quickly, reducing distortion
- Perfect for quick cuts for non-critical applications or in "hard to get at" places