Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Replacement Cables
Replacement Cables
Supplier:   Restek
Replacement Cables
Supplier:  Restek
Certificates
Replacement Cables
Replacement Cables
2 Options Available Below
Product Details & Documents
These are replacement cables for Agilent GCs, integrators and autosamplers.
Delivery information: Instructions and writing diagrams included.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • REST20652
    Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
    Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
    1,8 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • REST20657
    Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
    Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
    1,8 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items