O-rings, graphite
Supplier: Restek
Certificates
About this item
These O-rings are made of graphite. Ideal for Bruker/Varian 1177 injectors.
- Maximum operating temperature 450 °C
- Can used with split or splitless liners
Frequently Bought Together
These O-rings are made of graphite. Ideal for Bruker/Varian 1177 injectors.