VWR®, UV spectroscopy bandwidth standards
Catalog # 84833.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100 mL
- Description:Bandwidth standard –toluene in hexane - ratio of 268,7 nm peak to 266,8 nm trough
- Seq:02
- Environmentally Preferable:
Specifications
About this item
Bandwidth standards.
Suitable for UV spectroscopy applications.
Packaging: Six permanently sealed UV cuvettes or a 100 ml amber bottle.