Specifications
- Antibody type:Primary
- Antigen name:UDP glucuronosyltransferase 1 family, polypeptide A9
- Antigen symbol:UGT1A9
- Clonality:Monoclonal
- Clone:Clone UGT1A9/1229
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Gene ID:554822
- Antigen synonyms:UDPGT 1-9|UGT1-09|Ugt1a9|UDP-glucuronosyltransferase 1A9 (UGT1A9)|UGT-1I|GNT1|UDP-glucuronosyltransferase 1-I|lugP4|UGT1.9|HLUGP4|UGT1*9|UGT1I|UDPGT
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
This antibody recognizes a protein of about 60 kDa, which is identified as human UGT1A9. It does not cross-react with the other UGT1A isoforms including UGT1A7, UGT1A8, and UGT1A10 and shows a high degree of specificity. UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This MAb binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC s). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. This MAb also binds to cell preparations of hepatocellular carcinoma biopsies or cell lines.
Type: Primary
Antigen: UGT1A9
Clonality: Monoclonal
Clone: UGT1A9/1229
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human