Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF640R) [clone: B22.1 B23.1]
 :  Biotium
 
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF640R) [clone: B22.1 B23.1]
 :  Biotium

 

  • Catalog No:
  • BNC401221-100
  • BNC401221-500
  • Antigen symbol:
  • KRT8 & KRT18
  • KRT8 & KRT18
  • Antigen name:
  • Cytokeratin 8/18
  • Cytokeratin 8/18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone B22.1 B23.1
  • Clone B22.1 B23.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 533782 , 708445 (CK8); 406013 (CK18)
  • 533782 , 708445 (CK8); 406013 (CK18)
  • Isotype:
  • IgGs
  • IgGs
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together