Anti-NRD1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14048C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Nardilysin
- Antigen symbol:NRD1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:NRDC
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NRD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human