Anti-DPAGT1 Rabbit Polyclonal Antibody
ABGEAP13970A
: Abgent
- Antibody type:Primary
- Antigen symbol:DPAGT1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: DPAGT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human