About this item
This antibody recognizes a cell surface glycoprotein of 95/115/135 kDa (depending upon the extent of glycosylation), identified as CD43. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combination with anti-CD20, effective immunophenotyping of the lymphomas in formalin-fixed tissues can be obtained. Co-staining of a lymphoid infiltrate with anti-CD20 and anti-CD43 argues against a reactive process and favors a diagnosis of lymphoma.
Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: SPN/1094
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC401094-500
- BNC401094-100
- Antigen symbol:
- SPN
- SPN
- Antigen name:
- CD43
- CD43
- Antigen synonyms:
- LSN|Sialophorin|Leukosialin|SPN|Galactoglycoprotein|GPL115|GALGP|Leukocyte sialoglycoprotein
- LSN|Sialophorin|Leukosialin|SPN|Galactoglycoprotein|GPL115|GALGP|Leukocyte sialoglycoprotein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone SPN/1094
- Clone SPN/1094
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 632188
- 632188
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide