Order Entry
Slovenia
ContactUsLinkComponent
Anti-CR2 Mouse Monoclonal Antibody (CF405S) [clone: FR5A10]
Anti-CR2 Mouse Monoclonal Antibody (CF405S) [clone: FR5A10]
Supplier:  Biotium
Certificates
Anti-CR2 Mouse Monoclonal Antibody (CF405S) [clone: FR5A10]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041087-100
  • BNC041087-500
  • Antigen symbol:
  • CR2
  • CR2
  • Antigen name:
  • CD21
  • CD21
  • Antigen synonyms:
  • SLEB9|CVID7|Epstein-Barr virus receptor|EVBR|EBV receptor|CD21|Complement Receptor type 2 (CR2)|Complement C3d receptor (C3DR)|EBV-R
  • SLEB9|CVID7|Epstein-Barr virus receptor|EVBR|EBV receptor|CD21|Complement Receptor type 2 (CR2)|Complement C3d receptor (C3DR)|EBV-R
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone FR5A10
  • Clone FR5A10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 445757
  • 445757
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together