About this item
Recognizes a sialoglycoprotein of 39 kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation.
Type: Primary
Antigen: GYPA
Clonality: Monoclonal
Clone: A84-B/H2
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human
Specifications
- Catalog No:
- BNC401085-100
- BNC401085-500
- Antigen symbol:
- GYPA
- GYPA
- Antigen name:
- Glycophorin A
- Glycophorin A
- Antigen synonyms:
- GPA|GYPA|Sialoglycoprotein alpha|MNS|PAS2|Blood group--MN locus|MN sialoglycoprotein|GPErik|GpMiIII|GPSAT
- GPA|GYPA|Sialoglycoprotein alpha|MNS|PAS2|Blood group--MN locus|MN sialoglycoprotein|GPErik|GpMiIII|GPSAT
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone A84-B/H2
- Clone A84-B/H2
- Reactivity:
- Human, Bovine
- Human, Bovine
- Host:
- Mouse
- Mouse
- Gene ID:
- 434973 , 654368
- 434973 , 654368
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide