Order Entry
Slovenia
ContactUsLinkComponent
Anti-CDH16 Mouse Monoclonal Antibody (CF405S) [clone: CDH16/1071]
Anti-CDH16 Mouse Monoclonal Antibody (CF405S) [clone: CDH16/1071]
Supplier:  Biotium
Certificates
Anti-CDH16 Mouse Monoclonal Antibody (CF405S) [clone: CDH16/1071]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041071-500
  • BNC041071-100
  • Antigen symbol:
  • CDH16
  • CDH16
  • Antigen name:
  • Ksp-Cadherin / CDH16
  • Ksp-Cadherin / CDH16
  • Antigen synonyms:
  • Cadherin-16 (CDH16)|Ksp-cadherin antibody|Kidney-specific cadherin
  • Cadherin-16 (CDH16)|Ksp-cadherin antibody|Kidney-specific cadherin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone CDH16/1071
  • Clone CDH16/1071
  • Reactivity:
  • Rabbit, Human, Dog, Mouse
  • Rabbit, Human, Dog, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 513660
  • 513660
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together