Order Entry
Slovenia
ContactUsLinkComponent
Anti-TRIM29 Mouse Monoclonal Antibody (CF488A) [clone: TRIM29/1041]
Anti-TRIM29 Mouse Monoclonal Antibody (CF488A) [clone: TRIM29/1041]
Supplier:  Biotium
Certificates
Anti-TRIM29 Mouse Monoclonal Antibody (CF488A) [clone: TRIM29/1041]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881041-500
  • BNC881041-100
  • Antigen symbol:
  • TRIM29
  • TRIM29
  • Antigen name:
  • Tripartite Motif Containing 29
  • Tripartite Motif Containing 29
  • Antigen synonyms:
  • Tripartite motif-containing protein 29 (TRIM29)|Ataxia telangiectasia group D complementing gene (ATDC)
  • Tripartite motif-containing protein 29 (TRIM29)|Ataxia telangiectasia group D complementing gene (ATDC)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone TRIM29/1041
  • Clone TRIM29/1041
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 504115
  • 504115
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together