Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD70 Mouse Monoclonal Antibody (CF647) [clone: TNFS7/1026]
Anti-CD70 Mouse Monoclonal Antibody (CF647) [clone: TNFS7/1026]
Supplier:  Biotium
Certificates
Anti-CD70 Mouse Monoclonal Antibody (CF647) [clone: TNFS7/1026]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC471026-100
  • BNC471026-500
  • Antigen symbol:
  • CD70
  • CD70
  • Antigen synonyms:
  • Tumor necrosis factor (ligand) superfamily|member 7 (TNFSF7)|CD27LG|CD27 ligand (CD27L)|Ki24|Surface antigen CD70
  • Tumor necrosis factor (ligand) superfamily|member 7 (TNFSF7)|CD27LG|CD27 ligand (CD27L)|Ki24|Surface antigen CD70
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone TNFS7/1026
  • Clone TNFS7/1026
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 501497 , 715224
  • 501497 , 715224
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together