Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-FUT3 Mouse Monoclonal Antibody (CF405S) [clone: 2-25LE]
 :  Biotium
 
Anti-FUT3 Mouse Monoclonal Antibody (CF405S) [clone: 2-25LE]
 :  Biotium

 

  • Catalog No:
  • BNC041008-500
  • BNC041008-100
  • Antigen symbol:
  • FUT3
  • FUT3
  • Antigen name:
  • Lewis B
  • Lewis B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone 2-25LE
  • Clone 2-25LE
  • Reactivity:
  • Pig, Human, Guinea pig
  • Pig, Human, Guinea pig
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 169238
  • 169238
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together