Anti-HIF-PH2 Mouse Monoclonal Antibody [clone: 366G/76/3] (Biotin)
Catalog # NOVUNBP1-30328B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:HIF Prolyl Hydroxylase 2
- Antigen symbol:HIF-PH2
- Clonality:Monoclonal
- Clone:366G/76/3
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:54583
- Antigen synonyms:hypoxia-inducible factor prolyl hydroxylase 2|HPH-2|egl nine homolog 1|PNAS-118|PHD2|egl nine-like protein 1|HIF prolyl hydroxylase 2|HIFPH2|HIF-PH2|prolyl hydroxylase domain-containing protein 2|ECYT3|HIF-prolyl hydroxylase 2|C1orf12|ZMYND6|HPH2|zinc finger MYND domain-containing protein 6|SM20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Mouse Monoclonal HIF Prolyl Hydroxylase 2 Antibody (366G/76/3) [Biotin]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Type: Primary
Antigen: HIF-PH2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human