Anti-CHOP/GADD153 Mouse Monoclonal Antibody [clone: 9C8]
Supplier: Novus Biologicals
Certificates
About this item
Mouse Monoclonal CHOP/GADD153 Antibody (9C8). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: CHOP/GADD153
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- NOVUNB600-1335
- NOVUNB600-1335SS
- Antigen symbol:
- CHOP/GADD153
- CHOP/GADD153
- Antigen name:
- DNA-damage-inducible transcript 3
- DNA-damage-inducible transcript 3
- Antigen synonyms:
- GADD153|CHOP10|CHOP|CEBPZ|CHOP-10
- GADD153|CHOP10|CHOP|CEBPZ|CHOP-10
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9C8
- 9C8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1649
- 1649
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel shift:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together