Order Entry
Slovenia
ContactUsLinkComponent
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: DSD/958]
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: DSD/958]
Catalog # BNUM0958-50
Supplier:  Biotium
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: DSD/958]
Catalog # BNUM0958-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Double Stranded DNA
  • Clonality:
    Monoclonal
  • Clone:
    Clone DSD/958
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Human
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

This MAb recognizes the double stranded DNA in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in human cells. This MAb produces a homogeneous staining pattern in the nucleus of normal and malignant cells. Deoxyribonucleic acid (DNA) is a nucleic acid that stores long-term information regarding the development and function of all known living organisms. DNA consists of two long nucleotide polymers, which are composed of four bases, namely adenine, thymine, guanine and cytosine, all of which are flanked by a phosphate-deoxyribose backbone. Normally, DNA exists as a double-stranded (ds) molecule that forms in the shape of a double helix, allowing the bases and the backbone of the two strands to interact, thus forming a polynucleotide. When the double helix is unwound (either by enzymes or heat), DNA exists as a single-stranded (ss) molecule that is less stable than the double helix, but is necessary for protein access to DNA bases. Double stranded DNA markers are useful tools in biology research and aid in the study of DNA behavior and characteristics.

  • Purified, BSA-free

Type: Primary
Antigen: Double Stranded DNA
Clonality: Monoclonal
Clone: DSD/958
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human