Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-GYPA Mouse Monoclonal Antibody (Purified) [clone: A63-B/C2]
 :  Biotium
 
Anti-GYPA Mouse Monoclonal Antibody (Purified) [clone: A63-B/C2]
 :  Biotium

 

  • Catalog No:
  • BTIUBNUB0935-500
  • BTIUBNUB0935-100
  • Antigen symbol:
  • GYPA
  • GYPA
  • Antigen name:
  • Glycophorin A
  • Glycophorin A
  • Antigen synonyms:
  • GPA|GYPA|Sialoglycoprotein alpha|MNS|PAS2|Blood group--MN locus|MN sialoglycoprotein|GPErik|GpMiIII|GPSAT
  • GPA|GYPA|Sialoglycoprotein alpha|MNS|PAS2|Blood group--MN locus|MN sialoglycoprotein|GPErik|GpMiIII|GPSAT
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone A63-B/C2
  • Clone A63-B/C2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 434973 , 654368
  • 434973 , 654368
  • Isotype:
  • IgM kappa
  • IgM kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Frequently Bought Together