Order Entry
Slovenia
ContactUsLinkComponent
Anti-TYR Mouse Monoclonal Antibody (CF647) [clone: T311 OCA1/812]
Anti-TYR Mouse Monoclonal Antibody (CF647) [clone: T311 OCA1/812]
Supplier:  Biotium
Certificates
Anti-TYR Mouse Monoclonal Antibody (CF647) [clone: T311 OCA1/812]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470908-500
  • BNC470908-100
  • Antigen symbol:
  • TYR
  • TYR
  • Antigen name:
  • Tyrosinase
  • Tyrosinase
  • Antigen synonyms:
  • Tumor rejection antigen AB|ATN|CMM8|SHEP3|Oculocutaneous albinism IA|Monophenol monooxygenase|OCA1A|OCA1|TYR|LB24-AB|SK29-AB
  • Tumor rejection antigen AB|ATN|CMM8|SHEP3|Oculocutaneous albinism IA|Monophenol monooxygenase|OCA1A|OCA1|TYR|LB24-AB|SK29-AB
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone T311 OCA1/812
  • Clone T311 OCA1/812
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 503555
  • 503555
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together