Order Entry
Slovenia
ContactUsLinkComponent
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: SMMS-1]
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: SMMS-1]
Supplier:  Biotium
Certificates
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: SMMS-1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0893-500
  • BNCB0893-100
  • Antigen symbol:
  • MYH11
  • MYH11
  • Antigen name:
  • Myosin Smooth Muscle Heavy Chain
  • Myosin Smooth Muscle Heavy Chain
  • Antigen synonyms:
  • SMMHC|AAT4|Myosin heavy chain 11 smooth muscle|SM2|Smooth muscle myosin heavy chain 11|Myosin-11|Myosin heavy chain 11 (MYH11)|SM1|SMHC
  • SMMHC|AAT4|Myosin heavy chain 11 smooth muscle|SM2|Smooth muscle myosin heavy chain 11|Myosin-11|Myosin heavy chain 11 (MYH11)|SM1|SMHC
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone SMMS-1
  • Clone SMMS-1
  • Reactivity:
  • Pig, Rabbit, Chicken, Human, Bovine, Guinea pig, Dog, Rat, Cat
  • Pig, Rabbit, Chicken, Human, Bovine, Guinea pig, Dog, Rat, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 460109
  • 460109
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together