Anti-Ep-CAM Mouse Monoclonal Antibody (Biotin) [clone: EGP40/837]
Supplier: Biotium
Certificates
About this item
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas with the exception of adult squamous epithelium, hepatocytes and gastric epithelial cells. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.
Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: EGP40/837
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNCB0837-500
- BNCB0837-100
- Antigen symbol:
- EPCAM
- EPCAM
- Antigen name:
- Epithelial cell adhesion molecule
- Epithelial cell adhesion molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone EGP40/837
- Clone EGP40/837
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 542050
- 542050
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together