Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-CALD1 Mouse Monoclonal Antibody (CF568) [clone: h-CALD]
 :  Biotium
 
Anti-CALD1 Mouse Monoclonal Antibody (CF568) [clone: h-CALD]
 :  Biotium

 

  • Catalog No:
  • BNC680819-100
  • BNC680819-500
  • Antigen symbol:
  • CALD1
  • CALD1
  • Antigen name:
  • Caldesmon HMW
  • Caldesmon HMW
  • Antigen synonyms:
  • HCAD|Caldesmon 1 Isoform 1|Caldesmon 1 Isoform 5|NAG22|CALD1|Caldesmon 1 Isoform 2|Caldesmon 1 Isoform 4|CDM|CAD|Caldesmon 1 Isoform 3|LCAD
  • HCAD|Caldesmon 1 Isoform 1|Caldesmon 1 Isoform 5|NAG22|CALD1|Caldesmon 1 Isoform 2|Caldesmon 1 Isoform 4|CDM|CAD|Caldesmon 1 Isoform 3|LCAD
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone h-CALD
  • Clone h-CALD
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 490203
  • 490203
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together