Order Entry
Slovenia
ContactUsLinkComponent
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF647) [clone: PY793]
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF647) [clone: PY793]
Supplier:  Biotium
Certificates
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF647) [clone: PY793]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470793-500
  • BNC470793-100
  • Antigen symbol:
  • pY
  • pY
  • Antigen name:
  • Phosphotyrosine
  • Phosphotyrosine
  • Antigen synonyms:
  • P-Tyr
  • P-Tyr
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone PY793
  • Clone PY793
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • lgG2b
  • lgG2b
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together