Order Entry
Slovenia
ContactUsLinkComponent
Anti-ERBB2 Mouse Monoclonal Antibody (Biotin) [clone: ERB2/776]
Anti-ERBB2 Mouse Monoclonal Antibody (Biotin) [clone: ERB2/776]
Supplier:  Biotium
Certificates
Anti-ERBB2 Mouse Monoclonal Antibody (Biotin) [clone: ERB2/776]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0776-500
  • BNCB0776-100
  • Antigen symbol:
  • ERBB2
  • ERBB2
  • Antigen name:
  • HER-2 / CD340
  • HER-2 / CD340
  • Antigen synonyms:
  • Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
  • Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone ERB2/776
  • Clone ERB2/776
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 446352
  • 446352
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together