Order Entry
Slovenia
ContactUsLinkComponent
Anti-HSPB1 Mouse Monoclonal Antibody (CF405S) [clone: HSPB1/774]
Anti-HSPB1 Mouse Monoclonal Antibody (CF405S) [clone: HSPB1/774]
Supplier:  Biotium
Certificates
Anti-HSPB1 Mouse Monoclonal Antibody (CF405S) [clone: HSPB1/774]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BTIUBNC040774-500
  • BTIUBNC040774-100
  • Antigen symbol:
  • HSPB1
  • HSPB1
  • Antigen name:
  • HSP27
  • HSP27
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone HSPB1/774
  • Clone HSPB1/774
  • Reactivity:
  • Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
  • Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 520973
  • 520973
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together