Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD8A Mouse Monoclonal Antibody [clone: C8/468 C8/144B]
Anti-CD8A Mouse Monoclonal Antibody [clone: C8/468 C8/144B]
Catalog # BNUM0750-50
Supplier:  Biotium
Anti-CD8A Mouse Monoclonal Antibody [clone: C8/468 C8/144B]
Catalog # BNUM0750-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    Clone C8/468 C8/144B
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgGs
  • Reactivity:
    Human
  • Gene ID:
    85258
  • Antigen synonyms:
    T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4 /CD8-. Anaplastic large cell lymphoma is usually CD4 and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.

  • Purified
  • BSA-free

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: C8/468 C8/144B
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human