Order Entry
Slovenia
ContactUsLinkComponent
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: GP1.4 E29]
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: GP1.4 E29]
Supplier:  Biotium
Certificates
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: GP1.4 E29]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940743-500
  • BNC940743-100
  • Antigen symbol:
  • MUC1
  • MUC1
  • Antigen name:
  • Mucin 1 / EMA / Episialin / CD227
  • Mucin 1 / EMA / Episialin / CD227
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone GP1.4 E29
  • Clone GP1.4 E29
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 89603
  • 89603
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together