Order Entry
Slovenia
ContactUsLinkComponent
Anti-MLANA Mouse Monoclonal Antibody (CF488A) [clone: HMB45 A103 T311]
Anti-MLANA Mouse Monoclonal Antibody (CF488A) [clone: HMB45 A103 T311]
Supplier:  Biotium
Certificates
Anti-MLANA Mouse Monoclonal Antibody (CF488A) [clone: HMB45 A103 T311]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880702-500
  • BNC880702-100
  • Antigen symbol:
  • MLANA
  • MLANA
  • Antigen name:
  • Melanoma Marker
  • Melanoma Marker
  • Antigen synonyms:
  • MLAN-A|Melanoma antigen recognized by T-cells 1 (MART-1)|TYR|PMEL17
  • MLAN-A|Melanoma antigen recognized by T-cells 1 (MART-1)|TYR|PMEL17
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone HMB45 A103 T311
  • Clone HMB45 A103 T311
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 154069-MART-1; 503555-Tyrosinase; 95972-gp100
  • 154069-MART-1; 503555-Tyrosinase; 95972-gp100
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together