Recognizes a cell surface protein of 220-240 kDa, identified as L1 cell adhesion molecule. The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, and adducted thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, and neural migration as well as in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising from neuroectoderm. This MAb is useful in the identification of primitive neuroectodermal tumors. It binds to tumors of neuroectodermal and glial origin e.g. neuroblastoma and Schwannomas. It does not bind to pediatric or adult brain.
Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: UJ127
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNC400679-100
- BNC400679-500
- Antigen symbol:
- L1CAM
- L1CAM
- Antigen name:
- CD171 / L1CAM
- CD171 / L1CAM
- Antigen synonyms:
- L1 Cell Adhesion Molecule|Hyd|HSAS1|CAML1|S10|SPG1|NILE|Neural cell adhesion molecule L1|L1-NCAM|MASA|Nerve-growth factor-inducible large external glycoprotein|NCAM-L1|MIC5
- L1 Cell Adhesion Molecule|Hyd|HSAS1|CAML1|S10|SPG1|NILE|Neural cell adhesion molecule L1|L1-NCAM|MASA|Nerve-growth factor-inducible large external glycoprotein|NCAM-L1|MIC5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone UJ127
- Clone UJ127
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 52818
- 52818
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide