Order Entry
Slovenia
ContactUsLinkComponent
Anti-L1CAM Mouse Monoclonal Antibody (CF568) [clone: UJ127]
Anti-L1CAM Mouse Monoclonal Antibody (CF568) [clone: UJ127]
Supplier:  Biotium
Certificates
Anti-L1CAM Mouse Monoclonal Antibody (CF568) [clone: UJ127]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680679-100
  • BNC680679-500
  • Antigen symbol:
  • L1CAM
  • L1CAM
  • Antigen name:
  • CD171 / L1CAM
  • CD171 / L1CAM
  • Antigen synonyms:
  • L1 Cell Adhesion Molecule|Hyd|HSAS1|CAML1|S10|SPG1|NILE|Neural cell adhesion molecule L1|L1-NCAM|MASA|Nerve-growth factor-inducible large external glycoprotein|NCAM-L1|MIC5
  • L1 Cell Adhesion Molecule|Hyd|HSAS1|CAML1|S10|SPG1|NILE|Neural cell adhesion molecule L1|L1-NCAM|MASA|Nerve-growth factor-inducible large external glycoprotein|NCAM-L1|MIC5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone UJ127
  • Clone UJ127
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 52818
  • 52818
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together