Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody (CF647) [clone: LPSR/654]
Anti-CD14 Mouse Monoclonal Antibody (CF647) [clone: LPSR/654]
Supplier:  Biotium
Certificates
Anti-CD14 Mouse Monoclonal Antibody (CF647) [clone: LPSR/654]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470654-100
  • BNC470654-500
  • Antigen symbol:
  • CD14
  • CD14
  • Antigen synonyms:
  • Monocyte Differentiation Antigen 14|LPSR|Mo2|Myeloid cell-specific leucine-rich glycoprotein|LPS-R|Lipopolysaccharide receptor
  • Monocyte Differentiation Antigen 14|LPSR|Mo2|Myeloid cell-specific leucine-rich glycoprotein|LPS-R|Lipopolysaccharide receptor
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone LPSR/654
  • Clone LPSR/654
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 163867
  • 163867
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together