Order Entry
Slovenia
ContactUsLinkComponent
Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: VWF635]
Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: VWF635]
Supplier:  Biotium
Certificates
Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: VWF635]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880635-100
  • BNC880635-500
  • Antigen symbol:
  • VWF
  • VWF
  • Antigen name:
  • von Willebrand Factor
  • von Willebrand Factor
  • Antigen synonyms:
  • F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
  • F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone VWF635
  • Clone VWF635
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 440848
  • 440848
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together