Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD63 Mouse Monoclonal Antibody (CF647) [clone: MX-49.129.5]
Anti-CD63 Mouse Monoclonal Antibody (CF647) [clone: MX-49.129.5]
Supplier:  Biotium
Certificates
Anti-CD63 Mouse Monoclonal Antibody (CF647) [clone: MX-49.129.5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470525-500
  • BNC470525-100
  • Antigen symbol:
  • CD63
  • CD63
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone MX-49.129.5
  • Clone MX-49.129.5
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 445570
  • 445570
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together