Order Entry
Slovenia
ContactUsLinkComponent
Anti-PAX7 Mouse Monoclonal Antibody (CF640R) [clone: PAX7/497]
Anti-PAX7 Mouse Monoclonal Antibody (CF640R) [clone: PAX7/497]
Supplier:  Biotium
Certificates
Anti-PAX7 Mouse Monoclonal Antibody (CF640R) [clone: PAX7/497]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400497-500
  • BNC400497-100
  • Antigen symbol:
  • PAX7
  • PAX7
  • Antigen name:
  • Paired box 7
  • Paired box 7
  • Antigen synonyms:
  • PAX7 transcriptional factor|RMS2|Paired box gene 7|HUP1|PAX7B|Paired domain gene HuP1|Paired box homeotic gene 7|PAX7/FKHR fusion gene
  • PAX7 transcriptional factor|RMS2|Paired box gene 7|HUP1|PAX7B|Paired domain gene HuP1|Paired box homeotic gene 7|PAX7/FKHR fusion gene
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone PAX7/497
  • Clone PAX7/497
  • Reactivity:
  • Zebrafish, Chicken, Human, Rat, Mouse
  • Zebrafish, Chicken, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 113253
  • 113253
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together