About this item
The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma).
Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7/497
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Human, Mouse, Rat, Zebrafish
Specifications
- Catalog No:
- BNC940497-500
- BNC940497-100
- Antigen symbol:
- PAX7
- PAX7
- Antigen name:
- Paired box 7
- Paired box 7
- Antigen synonyms:
- PAX7 transcriptional factor|RMS2|Paired box gene 7|HUP1|PAX7B|Paired domain gene HuP1|Paired box homeotic gene 7|PAX7/FKHR fusion gene
- PAX7 transcriptional factor|RMS2|Paired box gene 7|HUP1|PAX7B|Paired domain gene HuP1|Paired box homeotic gene 7|PAX7/FKHR fusion gene
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone PAX7/497
- Clone PAX7/497
- Reactivity:
- Zebrafish, Chicken, Human, Rat, Mouse
- Zebrafish, Chicken, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 113253
- 113253
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide