Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-TOP1MT Mouse Monoclonal Antibody (CF405S) [clone: TOP1MT/488]
 :  Biotium
 
Anti-TOP1MT Mouse Monoclonal Antibody (CF405S) [clone: TOP1MT/488]
 :  Biotium

 

  • Catalog No:
  • BNC040488-500
  • BNC040488-100
  • Antigen symbol:
  • TOP1MT
  • TOP1MT
  • Antigen name:
  • Topoisomerase I MT
  • Topoisomerase I MT
  • Antigen synonyms:
  • Type IB Topoisomerase|Topoisomerase I mitochondrial|TOP1MT|DNA topoisomerase I
  • Type IB Topoisomerase|Topoisomerase I mitochondrial|TOP1MT|DNA topoisomerase I
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone TOP1MT/488
  • Clone TOP1MT/488
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 528574
  • 528574
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together