Order Entry
Slovenia
ContactUsLinkComponent
Anti-UPK3A Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]
Anti-UPK3A Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]
Supplier:  Biotium
Certificates
Anti-UPK3A Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940409-500
  • BNC940409-100
  • Antigen symbol:
  • UPK3A
  • UPK3A
  • Antigen name:
  • Uroplakin 3A
  • Uroplakin 3A
  • Antigen synonyms:
  • UPIIIA|Uroplakin3A|UPK3A|Uroplakin III (UPIII)|UP3a
  • UPIIIA|Uroplakin3A|UPK3A|Uroplakin III (UPIII)|UP3a
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone Rabbit PAb
  • Clone Rabbit PAb
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 632787
  • 632787
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together