Anti-UPK3A Rabbit Polyclonal Antibody (Biotin) [clone: Rabbit PAb]
Supplier: Biotium
Certificates
About this item
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.
Type: Primary
Antigen: UPK3A
Clonality: Polyclonal
Clone: Rabbit PAb
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- BNCB0409-500
- BNCB0409-100
- Antigen symbol:
- UPK3A
- UPK3A
- Antigen name:
- Uroplakin 3A
- Uroplakin 3A
- Antigen synonyms:
- UPIIIA|Uroplakin3A|UPK3A|Uroplakin III (UPIII)|UP3a
- UPIIIA|Uroplakin3A|UPK3A|Uroplakin III (UPIII)|UP3a
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone Rabbit PAb
- Clone Rabbit PAb
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 632787
- 632787
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together