About this item
The mouse monoclonal antibody recognizes CD1b, a 44 kDa type I glycoprotein associated with beta2-microglobulin. It is expressed on dendritic cells, Langerhans cells, thymocytes, and T acute lymphoblastic leukemia cells. The CD1 multigene family encodes five forms of the CD1 T-cell surface glycoprotein in human, designated CD1A, 1B, 1C, 1D and 1E. CD1, a type 1 membrane protein, has structural similarity to the MHC class I antigen and has been shown to present lipid antigens for recognition by T lymphocytes. Constitutive endocytosis of CD1B molecules and the differential sorting of MHC class II from lysosomes separate peptide- and lipid antigen-presenting molecules during dendritic cell maturation. CD1B is also expressed in interdigitating cells.
Type: Primary
Antigen: CD1B
Clonality: Monoclonal
Clone: RIV12
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC880317-100
- BNC880317-500
- Antigen symbol:
- CD1B
- CD1B
- Antigen name:
- CD1b molecule
- CD1b molecule
- Antigen synonyms:
- A1 domain|Cortical thymocyte antigen CD1B|T6/Leu6|CD1B|Ly38|hTa1 thymocyte antigen|T cell surface glycoprotein CD1b
- A1 domain|Cortical thymocyte antigen CD1B|T6/Leu6|CD1B|Ly38|hTa1 thymocyte antigen|T cell surface glycoprotein CD1b
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone RIV12
- Clone RIV12
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1310
- 1310
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide