Order Entry
Slovenia
ContactUsLinkComponent
Anti-LAMC1 Rat Monoclonal Antibody (CF647) [clone: A5]
Anti-LAMC1 Rat Monoclonal Antibody (CF647) [clone: A5]
Supplier:  Biotium
Certificates
Anti-LAMC1 Rat Monoclonal Antibody (CF647) [clone: A5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470308-100
  • BNC470308-500
  • Antigen symbol:
  • LAMC1
  • LAMC1
  • Antigen name:
  • Laminin
  • Laminin
  • Antigen synonyms:
  • Laminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1
  • Laminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone A5
  • Clone A5
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 609663
  • 609663
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together