Anti-NKX2-2 Mouse Monoclonal Antibody (CF647) [clone: NX2/294]
Supplier: Biotium
Certificates
About this item
Expression of NKX2.2 has been found in neuroendocrine tumors of the gut, making it a potential marker for the study of gastrointestinal neuroendocrine tumors. More recently, NKX2.2 protein was identified as a target of EWS-FLI-1, the fusion protein specific to Ewing sarcoma, and was shown to be differentially upregulated in Ewing sarcoma on the basis of array-based gene expression analysis. It acts as a valuable marker for Ewing sarcoma, with a sensitivity of 93% and a specificity of 89%, and aids in the differential diagnosis of small round cell tumors.
Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: NX2/294
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Chicken, Human, Mouse, Rat
Specifications
- Catalog No:
- BNC470294-100
- BNC470294-500
- Antigen symbol:
- NKX2-2
- NKX2-2
- Antigen name:
- NK2 homeobox 2
- NK2 homeobox 2
- Antigen synonyms:
- NK2 transcription factor like protein B|NKX22|NK2 transcription factor related locus 2|Nkx2b|tinman|Homeobox protein NK-2 homolog B
- NK2 transcription factor like protein B|NKX22|NK2 transcription factor related locus 2|Nkx2b|tinman|Homeobox protein NK-2 homolog B
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone NX2/294
- Clone NX2/294
- Reactivity:
- Chicken, Human, Rat, Mouse
- Chicken, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 516922
- 516922
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together