Order Entry
Slovenia
ContactUsLinkComponent
Anti-IGKC Mouse Monoclonal Antibody (CF594) [clone: KLC264]
Anti-IGKC Mouse Monoclonal Antibody (CF594) [clone: KLC264]
Supplier:  Biotium
Certificates
Anti-IGKC Mouse Monoclonal Antibody (CF594) [clone: KLC264]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940264-500
  • BNC940264-100
  • Antigen symbol:
  • IGKC
  • IGKC
  • Antigen name:
  • Kappa Light Chain
  • Kappa Light Chain
  • Antigen synonyms:
  • Ig Kappa Chain C Region|IGKC|HCAK1|Immunoglobulin KM
  • Ig Kappa Chain C Region|IGKC|HCAK1|Immunoglobulin KM
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone KLC264
  • Clone KLC264
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 449609
  • 449609
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together