Anti-Testosterone Mouse Monoclonal Antibody (CF488A) [clone: 4E1G2]
Supplier: Biotium
Certificates
About this item
This MAb is highly specific to testosterone. Its affinity constant for testosterone is ~1010M-1. In competitive binding immunoassay, it reacts with testosterone 100%, 11-beta-hydroxy testosterone 3.3%, 17-alpha-methyl testosterone
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species
Specifications
- Catalog No:
- BNC880250-100
- BNC880250-500
- Antigen name:
- Testosterone
- Testosterone
- Antigen synonyms:
- 4-Androsten-17-beta-ol-3-one|trans-Testosterone|17-beta-Hydroxy-4-androsten-3-one|17-beta-Hydroxy-3-oxo-4-androstene
- 4-Androsten-17-beta-ol-3-one|trans-Testosterone|17-beta-Hydroxy-4-androsten-3-one|17-beta-Hydroxy-3-oxo-4-androstene
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone 4E1G2
- Clone 4E1G2
- Reactivity:
- Multiple
- Multiple
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together