Anti-CD27 Mouse Monoclonal Antibody (CF647) [clone: 203.6]
Supplier: Biotium
Certificates
About this item
Recognizes a protein of a disulfide-linked 120 kDa dimer, identified as CD27 (Workshop VI; Code 6T-028). CD27 is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell co-stimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFB and SAPK/JNK and induces apoptosis.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 203.6
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC470163-500
- BNC470163-100
- Antigen symbol:
- CD27
- CD27
- Antigen synonyms:
- Tp55|S152|T-cell activation antigen CD27|TNFSF7|Tumor necrosis factor receptor superfamily member 7|T14|TNFRSF7|T cell activation antigen S152|LPFS2
- Tp55|S152|T-cell activation antigen CD27|TNFSF7|Tumor necrosis factor receptor superfamily member 7|T14|TNFRSF7|T cell activation antigen S152|LPFS2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone 203.6
- Clone 203.6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 355307
- 355307
- Isotype:
- IgG3 kappa
- IgG3 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together