Order Entry
Slovenia
ContactUsLinkComponent
Anti-CDKN1A Mouse Monoclonal Antibody (CF568) [clone: WA-1]
Anti-CDKN1A Mouse Monoclonal Antibody (CF568) [clone: WA-1]
Supplier:  Biotium
Certificates
Anti-CDKN1A Mouse Monoclonal Antibody (CF568) [clone: WA-1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680043-100
  • BNC680043-500
  • Antigen symbol:
  • CDKN1A
  • CDKN1A
  • Antigen name:
  • Cyclin-dependent kinase inhibitor 1A
  • Cyclin-dependent kinase inhibitor 1A
  • Antigen synonyms:
  • p21|WAF1
  • p21|WAF1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone WA-1
  • Clone WA-1
  • Reactivity:
  • Monkey, Chimpanzee, Human, Rat, Mouse
  • Monkey, Chimpanzee, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 370771
  • 370771
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together