Order Entry
Slovenia
ContactUsLinkComponent
Anti-CR1 Mouse Monoclonal Antibody (CF640R) [clone: E11]
Anti-CR1 Mouse Monoclonal Antibody (CF640R) [clone: E11]
Supplier:  Biotium
Certificates
Anti-CR1 Mouse Monoclonal Antibody (CF640R) [clone: E11]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400040-500
  • BNC400040-100
  • Antigen symbol:
  • CR1
  • CR1
  • Antigen name:
  • CD35 / CR1
  • CD35 / CR1
  • Antigen synonyms:
  • C3BR|Complement Component (3b/4b) receptor 1 including Knops blood group system|C3b/C4b receptor|KN|C3 binding protein|Knops blood group antigen|Complement receptor type 1|C4BR
  • C3BR|Complement Component (3b/4b) receptor 1 including Knops blood group system|C3b/C4b receptor|KN|C3 binding protein|Knops blood group antigen|Complement receptor type 1|C4BR
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone E11
  • Clone E11
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 334019
  • 334019
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together